Learn More
Grainger Demolition Hammer 1 1/8 In Hex 35 Lb
Catalog No. 502749285
Encompass_Preferred
The Makita 14F120 Demolition Hammer Kit is a corded electric demolition hammer that delivers 13.5 Joules of impact energy and 1,100 BPM for heavy-duty chiseling, chipping and drilling in concrete. It has a powerful 12 Amp motor and accepts SDS-MAX bits up to 4-3/8" in length. The kit includes the hammer, bull point, cold chisel, clay spade and carrying case. This professional-grade demolition hammer is designed for the most demanding chiseling, chipping and drilling applications in concrete.
Catalog No. 50-274-9285 Supplier Grainger Supplier No. 14F120
The Makita 14F120 Demolition Hammer Kit is a corded electric demolition hammer that delivers 13.5 Joules of impact energy and 1,100 BPM for heavy-duty chiseling, chipping and drilling in concrete. It has a powerful 12 Amp motor and accepts SDS-MAX bits up to 4-3/8" in length. The kit includes the hammer, bull point, cold chisel, clay spade and carrying case. This professional-grade demolition hammer is designed for the most demanding chiseling, chipping and drilling applications in concrete.