Learn More
Grainger Demolotion Hammer SDS-MAX 11 Lb.
Catalog No. 502753401
Encompass_Preferred
The Makita 1-9/16 in. SDS-Max Type 1 Demolition Hammer Kit is a powerful, corded electric demolition hammer that delivers 2.5 ft-lbs of impact energy for heavy-duty chiseling, chipping and drilling in concrete and masonry. It has a variable speed trigger with speeds up to 1,100 BPM and accepts SDS-Max bits up to 4-3/8 in. long. The kit includes the hammer, chisel, bull point, flat chisel, and carrying case for portability. This demolition hammer is designed for the professional contractor who requires maximum power and performance.
Catalog No. 50-275-3401 Supplier Grainger Supplier No. 14F116
The Makita 1-9/16 in. SDS-Max Type 1 Demolition Hammer Kit is a powerful, corded electric demolition hammer that delivers 2.5 ft-lbs of impact energy for heavy-duty chiseling, chipping and drilling in concrete and masonry. It has a variable speed trigger with speeds up to 1,100 BPM and accepts SDS-Max bits up to 4-3/8 in. long. The kit includes the hammer, chisel, bull point, flat chisel, and carrying case for portability. This demolition hammer is designed for the professional contractor who requires maximum power and performance.