Learn More
Grainger Demolotion Hammer SDS-MAX 20 Lb.
Catalog No. 502753402
Encompass_Preferred
The Grainger 14F117 is a Westward brand 1/4-inch drive socket adapter that converts a 1/4-inch square drive to a 3/8-inch square drive. It is made from chrome vanadium steel for durability and allows use of 1/4-inch drive tools with 3/8-inch drive socket accessories. This socket adapter measures 1-1/2 inches long and has a black oxide finish to resist corrosion. It is rated up to 20,000 psi working torque. The 14F117 socket adapter provides versatility when using 1/4-inch and 3/8-inch drive tools.
Catalog No. 50-275-3402 Supplier Grainger Supplier No. 14F117
The Grainger 14F117 is a Westward brand 1/4-inch drive socket adapter that converts a 1/4-inch square drive to a 3/8-inch square drive. It is made from chrome vanadium steel for durability and allows use of 1/4-inch drive tools with 3/8-inch drive socket accessories. This socket adapter measures 1-1/2 inches long and has a black oxide finish to resist corrosion. It is rated up to 20,000 psi working torque. The 14F117 socket adapter provides versatility when using 1/4-inch and 3/8-inch drive tools.