Learn More
Starna Cells Inc DEMOUNTABLE CUVETTE
SDP
Catalog No. NC9311101
Encompass_Preferred
Demountable Cuvette, Short path length, Demountable, open-ended, Spectrosil quartz, 2.5L x 12.5W x 45H, 0.004mL, One end open when assembled, Two polished windows
Catalog No. NC9311101 Supplier Starna Cells Inc Supplier No. 20/OQ0.01
Demountable Cuvette, Short path length, Demountable, open-ended, Spectrosil quartz, 2.5L x 12.5W x 45H, 0.004mL, One end open when assembled, Two polished windows