Learn More
Perkin Elmer US LLC Demountable Organics Quartz ESI ZipTorch for Avio 200/500
Catalog No. 502117301
Encompass_Preferred
This demountable quartz torch, with PVDF O-Ringfree Holder, is the standard torch for aqueous solutions. It is recommended for use with HF applications (using sapphire or Pt injectors) or when removing the injector for cleaning without dismounting the torch is preferred.
Catalog No. 50-211-7301 Supplier Perkin Elmer US LLC Supplier No. N0811935
This demountable quartz torch, with PVDF O-Ringfree Holder, is the standard torch for aqueous solutions. It is recommended for use with HF applications (using sapphire or Pt injectors) or when removing the injector for cleaning without dismounting the torch is preferred.