Learn More
Perkin Elmer US LLC Demountable Organics Quartz Torch - Orange/Black Mark
Catalog No. 502111342
Encompass_Preferred
This demountable quartz torch, with PVDF O-ringfree holder, is engineered for use with organics. It is recommended for HF applications with sapphire or platinum injectors, or when removing the injector for cleaning without dismounting the torch is preferred.
Catalog No. 50-211-1342 Supplier Perkin Elmer US LLC Supplier No. N8152596
This demountable quartz torch, with PVDF O-ringfree holder, is engineered for use with organics. It is recommended for HF applications with sapphire or platinum injectors, or when removing the injector for cleaning without dismounting the torch is preferred.