Learn More
Perkin Elmer US LLC Demountable Platinum Injector - 2.0 mm I.D., Gray
Catalog No. 502112136
Encompass_Preferred
Quartz injectors are used for volatile organic solvent introduction and are recommended for a wide range of non-HF applications. The Injector Light Shield Disk (N8152682) is included.
Catalog No. 50-211-2136 Supplier Perkin Elmer US LLC Supplier No. N8152429
Add to Cart
Add to Cart
Request a Quote
Quartz injectors are used for volatile organic solvent introduction and are recommended for a wide range of non-HF applications. The Injector Light Shield Disk (N8152682) is included.