Learn More
Perkin Elmer US LLC Demountable Quartz Injector - 2.0 mm I.D., Black
Catalog No. 502111900
Encompass_Preferred
Quartz injectors are used for volatile organic solvent introduction and are recommended for a wide range of non-HF applications. The Injector Light Shield Disk (N8152682) is included.
Catalog No. 50-211-1900 Supplier Perkin Elmer US LLC Supplier No. N8152376
Quartz injectors are used for volatile organic solvent introduction and are recommended for a wide range of non-HF applications. The Injector Light Shield Disk (N8152682) is included.