Learn More
Perkin Elmer US LLC Demountable Quartz Torch - Black Mark
Catalog No. 502112159
Encompass_Preferred
The demountable quartz torch uses a removable and replaceable injector. It is recommended for HF applications (sapphire or Pt injectors) or when the user prefers to remove the injector for cleaning without dismounting the torch. It's included in the Geological Sample Kit (N8150040) and the Productivity Configuration's Sample Introduction Kit (N8150036).
Catalog No. 50-211-2159 Supplier Perkin Elmer US LLC Supplier No. N8152379
The demountable quartz torch uses a removable and replaceable injector. It is recommended for HF applications (sapphire or Pt injectors) or when the user prefers to remove the injector for cleaning without dismounting the torch. It's included in the Geological Sample Kit (N8150040) and the Productivity Configuration's Sample Introduction Kit (N8150036).