Learn More
Perkin Elmer US LLC Demountable Sapphire Injector, 1.6 mm I.D.
Catalog No. 502104887
Encompass_Preferred
This Sapphire Injector Cassette Mount is designed for use with demountable torches in NexION 300/350 ICP-MS series instruments. The demountable design allows the injector to be removed and replaced efficiently without dismounting the torch. Standard Injectors have an O-ring free direct connection to Cyclonic and PFA spray chambers while O-ring free Injectors have an O-ring free seal that reduces contamination, makes it easy to clean and easy to insert.
Catalog No. 50-210-4887 Supplier Perkin Elmer US LLC Supplier No. N8145112
This Sapphire Injector Cassette Mount is designed for use with demountable torches in NexION 300/350 ICP-MS series instruments. The demountable design allows the injector to be removed and replaced efficiently without dismounting the torch. Standard Injectors have an O-ring free direct connection to Cyclonic and PFA spray chambers while O-ring free Injectors have an O-ring free seal that reduces contamination, makes it easy to clean and easy to insert.