Learn More
Perkin Elmer US LLC Demountable Silicon Nitride ESI ZipTorch for Avio 200/500
Catalog No. 502117300
Encompass_Preferred
This demountable quartz torch, with PVDF O-Ringfree Holder, is the standard torch for aqueous solutions. It is recommended for use with HF applications (using sapphire or Pt injectors) or when removing the injector for cleaning without dismounting the torch is preferred.
Catalog No. 50-211-7300 Supplier Perkin Elmer US LLC Supplier No. N0811913
This demountable quartz torch, with PVDF O-Ringfree Holder, is the standard torch for aqueous solutions. It is recommended for use with HF applications (using sapphire or Pt injectors) or when removing the injector for cleaning without dismounting the torch is preferred.