Learn More
Perkin Elmer US LLC Demountable UHP Quartz Torch - Gray Mark
Catalog No. 502112913
Encompass_Preferred
This demountable ultra pure quartz torch, marked grey, allows for the use of different injectors depending on the application. It is recommended for HF applications (sapphire or Pt injectors) or if removing the injector for cleaning without dismounting the torch is preferred. A PVDF O-ring free holder is included. This torch is included in the PFA-Platinum HF-Resistant Sample Introduction Kit (N8150033).
Catalog No. 50-211-2913 Supplier Perkin Elmer US LLC Supplier No. N8152448
This demountable ultra pure quartz torch, marked grey, allows for the use of different injectors depending on the application. It is recommended for HF applications (sapphire or Pt injectors) or if removing the injector for cleaning without dismounting the torch is preferred. A PVDF O-ring free holder is included. This torch is included in the PFA-Platinum HF-Resistant Sample Introduction Kit (N8150033).