Learn More
Cell Signaling Technology Demyelinating Disease Targets Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502051369
Encompass
The Demyelinating Disease Targets Antibody Sampler Kit provides an economical means of detecting protein components of the myelin sheath. This kit includes a comprehensive selection of primary antibodies to analyze the expression and localization of proteins associated with myelination and nodes of Ranvier. Each antibody is provided in sufficient quantity to perform two western blot experiments.
  • Includes antibodies against key structural myelin proteins including MBP, MAG, PLP1, and MOG
  • Contains markers for nodes of Ranvier-associated proteins including Caspr, Caspr2, and neurofascin
  • Validated for western blotting, immunofluorescence, immunohistochemistry, and immunoprecipitation
  • Includes β3-tubulin as an axonal control
  • Supplied in a stabilized glycerol-based buffer at pH 7.5 for optimal performance
Catalog No. 50-205-1369 Supplier Cell Signaling Technology Supplier No. 42135T
The Demyelinating Disease Targets Antibody Sampler Kit provides an economical means of detecting protein components of the myelin sheath. This kit includes a comprehensive selection of primary antibodies to analyze the expression and localization of proteins associated with myelination and nodes of Ranvier. Each antibody is provided in sufficient quantity to perform two western blot experiments.
  • Includes antibodies against key structural myelin proteins including MBP, MAG, PLP1, and MOG
  • Contains markers for nodes of Ranvier-associated proteins including Caspr, Caspr2, and neurofascin
  • Validated for western blotting, immunofluorescence, immunohistochemistry, and immunoprecipitation
  • Includes β3-tubulin as an axonal control
  • Supplied in a stabilized glycerol-based buffer at pH 7.5 for optimal performance