Learn More
Med Vet International Feral Cat and Small Mammal Den, J1393, Each
Catalog No. 5000495980
Encompass_Preferred
Designed to fit inside a standard cage this den provides a frightened and fractious animal a quiet 'hiding place'. Secure the animal inside by closing the porthole. The den, with the animal, can be removed from the cage for cleaning, treatment, or transporting. Reduces the risk of injury to the handler while providing a stress-free environment for the animal. Fits any cage larger than a 24" x 24". Virtually unbreakable beige plastic. Molded carrying handle on top. Measures 15"(D) x 11"(H) x 10"(W).
Catalog No. 50-004-95980 Supplier Med Vet International Supplier No. JOR1393
Designed to fit inside a standard cage this den provides a frightened and fractious animal a quiet 'hiding place'. Secure the animal inside by closing the porthole. The den, with the animal, can be removed from the cage for cleaning, treatment, or transporting. Reduces the risk of injury to the handler while providing a stress-free environment for the animal. Fits any cage larger than a 24" x 24". Virtually unbreakable beige plastic. Molded carrying handle on top. Measures 15"(D) x 11"(H) x 10"(W).