Learn More
C LECTA GMBH DENARASE 5000 kU, R&D Enzymes & Nucleases
Supplier:  C LECTA GMBH 208045000K
Encompass_Preferred
For process development of biologicals such as viral vectors and vaccines The enzyme cleaves all forms of DNA and RNA and enables a simpler more efficient downstream process - Purity 99% - Activity 250 U/ul - Formulation buffered aqueous glycerol solution- Storage -20C - Produced in conformity with ISO 9001 standard- Manufacturing free of animal-derived raw materials and antibiotics- Depending on your application 10-100 U/ml is needed - Net price excluding VAT and shipment costs
c-LEcta will ship the Products, as the case may be and at its sole discretion, either directly from Leipzig, Germany, or from c-LEcta’s third party service provider in the US. In the latter case, c-LEcta has already incurred all costs and fees related to the import of the Products into the US. Such costs and fees will be passed on by c-LEcta to the customer without any deductions or surcharges, if the Products are shipped from c-LEcta’s third party service provider in the US to the customer
Catalog No. 50-000-59957
Add to Cart
Request a Quote