Learn More
C LECTA GMBH DENARASE High Salt 1000 kU Enzymes & Nucleases
Supplier:  C LECTA GMBH 220021000K
Encompass_Preferred
The salt-tolerant enzyme cleaves all forms of DNA and RNA and enables a simpler more efficient downstream process at salt concentrations above 150 mM - Purity 98% - Activity 250 U/ul - Formulation buffered aqueous glycerol solution - Storage Temperature -20C - Produced under ISO 9001 standard- Manufactured without the use of antibiotics Triton X-100 and raw materials of animal origin- - Net price excluding VAT and shipment costs
c-LEcta will ship the Products, as the case may be and at its sole discretion, either directly from Leipzig, Germany, or from c-LEcta’s third party service provider in the US. In the latter case, c-LEcta has already incurred all costs and fees related to the import of the Products into the US. Such costs and fees will be passed on by c-LEcta to the customer without any deductions or surcharges, if the Products are shipped from c-LEcta’s third party service provider in the US to the customer
Catalog No. 50-000-59951