Learn More
Medchemexpress LLC Denatonium benzoate | 3734-33-6 | 99.9% | 1 ML
SDP
Catalog No. 5000412598
Encompass_Preferred
Denatonium benzoate is widely recognized as the most bitter compound, commonly employed as a deterrent by stimulating bitter taste receptors (TAS2Rs). Research indicates that it can enhance insulin secretion, exhibit anti-inflammatory effects, and potentially increase allergy susceptibility.
  • Potent bittering agent.
  • Stimulates bitter taste receptors (TAS2Rs).
  • Enhances insulin secretion.
  • Exhibits anti-inflammatory effects.
  • Increases allergy susceptibility.
Catalog No. 50-004-12598 Supplier Medchemexpress LLC Supplier No. HYB114610MM/1ML
Denatonium benzoate is widely recognized as the most bitter compound, commonly employed as a deterrent by stimulating bitter taste receptors (TAS2Rs). Research indicates that it can enhance insulin secretion, exhibit anti-inflammatory effects, and potentially increase allergy susceptibility.
  • Potent bittering agent.
  • Stimulates bitter taste receptors (TAS2Rs).
  • Enhances insulin secretion.
  • Exhibits anti-inflammatory effects.
  • Increases allergy susceptibility.