Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Denatonium (benzoate) (Standard) | 3734-33-6 | 99.92% | 25 MG
SDP

Catalog No. 5000413926
Encompass_Preferred

Denatonium (benzoate) (Standard) is the analytical standard of Denatonium (benzoate), intended for research and analytical applications. It is known as the most bitter compound, commonly used as an aversion agent to prevent inappropriate ingestion in products like denatured alcohol, antifreeze, nail biting prevention, respirator mask fit testing, animal repellents, liquid soaps, and shampoos. The compound is an analytical standard used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.

  • Appearance: Solid
  • Color: White to off-white
  • Molecular weight: 446.58
  • Formula: C₂₈H₃₄N₂O₃

Catalog No. 50-004-13926 Supplier Medchemexpress LLC Supplier No. 25MGHYB1146R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Denatonium (benzoate) (Standard) is the analytical standard of Denatonium (benzoate), intended for research and analytical applications. It is known as the most bitter compound, commonly used as an aversion agent to prevent inappropriate ingestion in products like denatured alcohol, antifreeze, nail biting prevention, respirator mask fit testing, animal repellents, liquid soaps, and shampoos. The compound is an analytical standard used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.

  • Appearance: Solid
  • Color: White to off-white
  • Molecular weight: 446.58
  • Formula: C₂₈H₃₄N₂O₃

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.