Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Denatonium (benzoate) (standard) | 3734-33-6 | MFCD00031578 | 99.8% | 446.58 | C28H34N2O3 | 10 MG
SDP

Catalog No. 5000296711
Encompass_Preferred

Denatonium benzoate is a bittering (aversive) agent used as an analytical standard and research reagent. The material is provided in a small solid quantity intended for laboratory research; consult the safety data sheet for handling and regulatory information.

  • High purity (99.8% by LCMS)
  • Chemical formula C28H34N2O3 and molecular weight 446.58
  • Commonly used as an aversive/bittering agent and analytical standard
  • Supplied as a 10 MG solid quantity for laboratory use
  • Reference CAS number 3734-33-6 for unambiguous identification

Catalog No. 50-002-96711 Supplier Medchemexpress LLC Supplier No. HYB1146R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Denatonium benzoate is a bittering (aversive) agent used as an analytical standard and research reagent. The material is provided in a small solid quantity intended for laboratory research; consult the safety data sheet for handling and regulatory information.

  • High purity (99.8% by LCMS)
  • Chemical formula C28H34N2O3 and molecular weight 446.58
  • Commonly used as an aversive/bittering agent and analytical standard
  • Supplied as a 10 MG solid quantity for laboratory use
  • Reference CAS number 3734-33-6 for unambiguous identification

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.