Learn More
National Diagnostics DENATURATION SOLUTION 1 LTDEN
SDP
Catalog No. 5089990110
Encompass
Denaturation Solution, Ready-to-use buffer solution of sodium chloride and sodium hydroxide designed for Southern and Northern Blotting, 1 LT
Catalog No. 50-899-90110 Supplier National Diagnostics Supplier No. EC8751LTR
Denaturation Solution, Ready-to-use buffer solution of sodium chloride and sodium hydroxide designed for Southern and Northern Blotting, 1 LT