Learn More
Sigma Aldrich Fine Chemicals Biosciences Denaturation Solution for
Catalog No. 502900858
Encompass
Sigmas denaturing SOLUTIONon is a strong base that denatures double-stranded DNA so that it can be efficiently transferred to nylon membrane and subsequently hybridized to labeled probe. When used with a depurination SOLUTIONon treatment results in the hydrolysis of the phosphodiester backbone at the sites of depurination allowing for more efficient transfer of large DNA in neutral Southern transfers.
Catalog No. 50-290-0858 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. N15314L
Add to Cart
Add to Cart
Sigmas denaturing SOLUTIONon is a strong base that denatures double-stranded DNA so that it can be efficiently transferred to nylon membrane and subsequently hybridized to labeled probe. When used with a depurination SOLUTIONon treatment results in the hydrolysis of the phosphodiester backbone at the sites of depurination allowing for more efficient transfer of large DNA in neutral Southern transfers.