Learn More
Denatured Alcohol, 100%, StatLab™
SDP
Catalog No. STL95305
Change view
Click to view available options
Quantity:
5 gal.
Catalog No. Quantity
STL95305 5 gal.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STL95305 Supplier SLMP Llc Dba Statlab Medical Supplier No. 95305
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
100% Denatured Alcohol requiring no TTB approval.
  • Specially denatured alcohol is ethanol based and suitable for use in clinical laboratory protocols
  • Ideal for processing and staining protocols that reference 100% alcohol, ethanol, or 100% ethanol in Histology and Cytology laboratories
  • Miscible with all clearing reagents
Specifications
Color Clear
Quantity 5 gal.