Learn More
Bioworld Denaturing Lysis Buffer, 500 mL
Catalog No. 50253022
Encompass_Preferred
Denaturing Lysis Buffer Lysis buffer for extracting proteins from cells. Technical Information:1X concentrationTris-HCL, pH 7.4 (50mM)SDS (2%)
Catalog No. 50-253-022 Supplier Bioworld Supplier No. 220400272
Denaturing Lysis Buffer Lysis buffer for extracting proteins from cells. Technical Information:1X concentrationTris-HCL, pH 7.4 (50mM)SDS (2%)