Learn More
Bioworld Denaturing Lysis Buffer, 250 mL
Catalog No. 50253021
Encompass_Preferred
Denaturing Lysis Buffer Lysis buffer for extracting proteins from cells. Technical Information:1X concentrationTris-HCL, pH 7.4 (50mM)SDS (2%)
Catalog No. 50-253-021 Supplier Bioworld Supplier No. 220400271
Denaturing Lysis Buffer Lysis buffer for extracting proteins from cells. Technical Information:1X concentrationTris-HCL, pH 7.4 (50mM)SDS (2%)