Learn More
Biocare Medical, LLC Denaturing Solution (Elution Step) - 100 ml
SDP
Catalog No. 5082821
Encompass_Preferred
Denaturing Solution is used as an elution step when using a double stain technique (Biocare's 7-step double stain procedure). This will ensure that the second staining protocol will not cross react with the first staining protocol. Solution A (yellow) is a concentrate that must be diluted with Solution B (red, dilution buffer) for proper elution. It is recommended that the Denaturing Solution be used at different dilutions depending on tissue fixation and the type of antigen retrieval method used.
Catalog No. 50-828-21 Supplier Biocare Medical, LLC Supplier No. DNS001L
Denaturing Solution is used as an elution step when using a double stain technique (Biocare's 7-step double stain procedure). This will ensure that the second staining protocol will not cross react with the first staining protocol. Solution A (yellow) is a concentrate that must be diluted with Solution B (red, dilution buffer) for proper elution. It is recommended that the Denaturing Solution be used at different dilutions depending on tissue fixation and the type of antigen retrieval method used.