Learn More
TARGETMOL CHEMICALS INC DENDROBINE 10MG
SDP
Catalog No. 502576635
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Dendrobine has a slight but demonstrable analgesic and antipyretic action. 2. Dendrobine produces moderate hyperglycemia diminishes cardiac activity in large doses lowers the blood pressure. 3. Dendrobine on the electrical activity and on amino acid-induced depolarizations of primary afferent terminals were tested on the frog isolated spinal cord. purity: 99%
Catalog No. 50-257-6635 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S170810MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Dendrobine has a slight but demonstrable analgesic and antipyretic action. 2. Dendrobine produces moderate hyperglycemia diminishes cardiac activity in large doses lowers the blood pressure. 3. Dendrobine on the electrical activity and on amino acid-induced depolarizations of primary afferent terminals were tested on the frog isolated spinal cord. purity: 99%