Learn More
Cayman Chemical Dendrobine, Purity: ≥95%, CAS: 2115-91-5, 10 mg
SDP
Catalog No. 502936182
Encompass_Preferred
A sesquiterpenoid with anticancer activity; decreases viability of A549 cells at 2.5-15 µg/ml; induces apoptosis in A549 cells at 1-10 µg/ml, as well as increases JNK phosphorylation and sensitizes A549 cells to cisplatin at 10 µg/ml; reduces tumor growth in an A549 mouse xenograft model at 50 mg/kg per day with an additive effect when used in combination with cisplatin
Catalog No. 50-293-6182 Supplier Cayman Chemical Supplier No. 2606710 MG
A sesquiterpenoid with anticancer activity; decreases viability of A549 cells at 2.5-15 µg/ml; induces apoptosis in A549 cells at 1-10 µg/ml, as well as increases JNK phosphorylation and sensitizes A549 cells to cisplatin at 10 µg/ml; reduces tumor growth in an A549 mouse xenograft model at 50 mg/kg per day with an additive effect when used in combination with cisplatin