Learn More
Cayman Chemical Dendrophenol, Purity: ≥98%, CAS: 108853-14-1, 10 mg
SDP
Catalog No. 502978499
Encompass_Preferred
A polyphenol with diverse biological activities; decreases viability of, and induces cell cycle arrest at the S phase in, HCT116 cells at 10 µM; inhibits the migration of MDA-MB-231 cells at 3 or 10 µM; inhibits LPS-induced increases in iNOS, COX-2, HIF-1α, and NF-κB levels and NF-κB nuclear translocation in RAW 264.7 macrophages at 100 µM; reduces tumor size without affecting body weight in an HCT116 mouse xenograft model at 50 or 100 mg/kg per day; decreases total lung weight in an MDA-MB-231 mouse xenograft model at 100 mg/kg per day
Catalog No. 50-297-8499 Supplier Cayman Chemical Supplier No. 4211510 MG
A polyphenol with diverse biological activities; decreases viability of, and induces cell cycle arrest at the S phase in, HCT116 cells at 10 µM; inhibits the migration of MDA-MB-231 cells at 3 or 10 µM; inhibits LPS-induced increases in iNOS, COX-2, HIF-1α, and NF-κB levels and NF-κB nuclear translocation in RAW 264.7 macrophages at 100 µM; reduces tumor size without affecting body weight in an HCT116 mouse xenograft model at 50 or 100 mg/kg per day; decreases total lung weight in an MDA-MB-231 mouse xenograft model at 100 mg/kg per day