Learn More
TARGETMOL CHEMICALS INC DENDROPHENOL 5MG
SDP
Catalog No. 502580012
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dendrophenol (Moscatilin) is a natural product isolated from the stem of Dendrobium loddigesii Rolfe act as a NF-(kappa)B inhibitor. Antineoplastic activity. purity: 98%
Catalog No. 50-258-0012 Supplier TARGETMOL CHEMICALS INC Supplier No. T3S17075MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dendrophenol (Moscatilin) is a natural product isolated from the stem of Dendrobium loddigesii Rolfe act as a NF-(kappa)B inhibitor. Antineoplastic activity. purity: 98%