Learn More
Enzo Life Sciences Dendrotoxin I (Dendroaspis p. polylepis), (purified) (140µg). CAS: 107950-33-4
SDP
Supplier:  Enzo Life Sciences BMLKC1630140
Encompass_Preferred
K+ channel blocker. Blocks Kv1.2 and Kv1.1 channels (IC50=0.13 and 3.1nM in oocytes respectively, with higher values for mammalian cells), as well as heteromultimeric channels comprised of Kv1.2 and Kv1.1 with other Kv1 isoforms. K+ channel blocker. Blocks Kv1.2 and Kv1.1 channels (IC50=0.13 and 3.1nM in oocytes respectively, with higher values for mammalian cells), as well as heteromultimeric channels comprised of Kv1.2 and Kv1.1 with other Kv1 isoforms. Purity: ≥98% (HPLC). Formulation: Lyophilized. Solubility: Soluble in aqueous buffers (pH 7.5). Long Term Storage: -20°C.
Catalog No. 50-201-1078