Learn More
Medchemexpress LLC Dendrotoxin-I TFA | 99.1% | 7149.24 | 100 UG
SDP
Catalog No. 5000399424
Encompass_Preferred
Dendrotoxin-I TFA (DTX-I TFA) is a potent K+ channel blocker with IC50s of 0.13-50 nM for voltage-gated potassium channel subunits KV1.1, KV1.2, and KV1.6. This neurotoxin demonstrates potential for cancer research and is intended for research use only.
  • Potent K+ channel blocker: Exhibits IC50s of 0.13-50 nM against voltage-gated potassium channel subunits KV1.1, KV1.2, and KV1.6.
  • Neurotoxin with cancer research potential: Implicated in studies exploring its use in cancer research.
  • In vivo activity: In an animal model using nude mice with MCF-7 cells, it showed a significant tumor growth inhibition effect when combined with hyperthermia.
Catalog No. 50-003-99424 Supplier Medchemexpress LLC Supplier No. HYP3055A100UG
Dendrotoxin-I TFA (DTX-I TFA) is a potent K+ channel blocker with IC50s of 0.13-50 nM for voltage-gated potassium channel subunits KV1.1, KV1.2, and KV1.6. This neurotoxin demonstrates potential for cancer research and is intended for research use only.
  • Potent K+ channel blocker: Exhibits IC50s of 0.13-50 nM against voltage-gated potassium channel subunits KV1.1, KV1.2, and KV1.6.
  • Neurotoxin with cancer research potential: Implicated in studies exploring its use in cancer research.
  • In vivo activity: In an animal model using nude mice with MCF-7 cells, it showed a significant tumor growth inhibition effect when combined with hyperthermia.