Learn More
Medchemexpress LLC Dendrotoxin-I (TFA) | 99.9% | 7149.24 | 5 MG
SDP
Catalog No. 5000398435
Encompass_Preferred
Dendrotoxin-I (DTX-I) TFA is a potent K+ channel blocker with IC50s of 0.13-50 nM for voltage-gated potassium channel subunits KV1.1, KV1.2, and KV1.6. This neurotoxin has potential for cancer research.
  • Potent K+ channel blocker.
  • Targets voltage-gated potassium channel subunits KV1.1, KV1.2, and KV1.6.
  • Potential for cancer research.
  • Shows significant tumor growth inhibition effect in nude mice with MCF-7 cells when combined with hyperthermia (at 5 mg/kg IV).
Catalog No. 50-003-98435 Supplier Medchemexpress LLC Supplier No. HYP3055A5MG
Dendrotoxin-I (DTX-I) TFA is a potent K+ channel blocker with IC50s of 0.13-50 nM for voltage-gated potassium channel subunits KV1.1, KV1.2, and KV1.6. This neurotoxin has potential for cancer research.
  • Potent K+ channel blocker.
  • Targets voltage-gated potassium channel subunits KV1.1, KV1.2, and KV1.6.
  • Potential for cancer research.
  • Shows significant tumor growth inhibition effect in nude mice with MCF-7 cells when combined with hyperthermia (at 5 mg/kg IV).