Learn More
Medchemexpress LLC Dendrotoxin K TFA | 119128-61-9 | 6559.66 | 500 UG
SDP
Catalog No. 5000398662
Encompass_Preferred
Dendrotoxin K functions as a Kv1.1 channel blocker. It plays a role in determining glutamate release within CA3 neurons by controlling the presynaptic spike waveform in a time-dependent manner.
  • Acts as a Kv1.1 channel blocker.
  • Influences glutamate release in CA3 neurons.
  • Regulates the presynaptic spike waveform in a time-dependent manner.
  • Intended for research use only.
Catalog No. 50-003-98662 Supplier Medchemexpress LLC Supplier No. HYP3089A500UG
Dendrotoxin K functions as a Kv1.1 channel blocker. It plays a role in determining glutamate release within CA3 neurons by controlling the presynaptic spike waveform in a time-dependent manner.
  • Acts as a Kv1.1 channel blocker.
  • Influences glutamate release in CA3 neurons.
  • Regulates the presynaptic spike waveform in a time-dependent manner.
  • Intended for research use only.