Learn More
Medchemexpress LLC Dendrotoxin K TFA | 99.5% | 6559.66 | 5 MG
SDP
Catalog No. 5000399407
Encompass_Preferred
Dendrotoxin K TFA is a Kv1.1 channel blocker. It determines glutamate release in CA3 neurons in a time-dependent manner through the control of the presynaptic spike waveform. The product is a solid, white to off-white in color.
  • Kv1.1 channel blocker
  • Determines glutamate release in CA3 neurons
  • Solid appearance
  • White to off-white color
Catalog No. 50-003-99407 Supplier Medchemexpress LLC Supplier No. HYP3089A5MG
Dendrotoxin K TFA is a Kv1.1 channel blocker. It determines glutamate release in CA3 neurons in a time-dependent manner through the control of the presynaptic spike waveform. The product is a solid, white to off-white in color.
  • Kv1.1 channel blocker
  • Determines glutamate release in CA3 neurons
  • Solid appearance
  • White to off-white color