Learn More
Medchemexpress LLC Dendrotoxin K TFA | 99.5% | 6559.66 (free base) | 1 MG
SDP
Catalog No. 5000396625
Encompass_Preferred
Dendrotoxin K TFA is a Kv1.1 channel blocker. It determines glutamate release in CA3 neurons in a time-dependent manner through the control of the presynaptic spike waveform.
  • Kv1.1 channel blocker
  • Determines glutamate release in CA3 neurons
  • Controls presynaptic spike waveform
Catalog No. 50-003-96625 Supplier Medchemexpress LLC Supplier No. HYP3089A1MG
Dendrotoxin K TFA is a Kv1.1 channel blocker. It determines glutamate release in CA3 neurons in a time-dependent manner through the control of the presynaptic spike waveform.
  • Kv1.1 channel blocker
  • Determines glutamate release in CA3 neurons
  • Controls presynaptic spike waveform