Learn More
Medchemexpress LLC Denfivontinib hydrochloride | 1457983-28-6 | MFCD28167815 | 98.0% | 521.41 g/mol | C25H25BrN6O2 | 10MM 1ML
SDP
Catalog No. 5000238105
Encompass_Preferred
Denfivontinib hydrochloride is a potent, ATP-competitive FLT3 inhibitor used in preclinical research on acute myeloid leukemia and agent resistance. The product is supplied as a 10 mM solution in DMSO (1 mL) and is intended for research use only. Chemical identifiers include CAS 1457983-28-6, formula C25H25BrN6O2, and molecular weight 521.41 g/mol.
  • Potent ATP-competitive FLT3 inhibitor.
  • Supplied as a 10 mM solution in DMSO (1 mL).
  • Suitable for preclinical AML and resistance research.
  • High purity (98.0%).
  • Provided as the hydrochloride salt form.
Catalog No. 50-002-38105 Supplier Medchemexpress LLC Supplier No. HY1233310MM 1ML
Denfivontinib hydrochloride is a potent, ATP-competitive FLT3 inhibitor used in preclinical research on acute myeloid leukemia and agent resistance. The product is supplied as a 10 mM solution in DMSO (1 mL) and is intended for research use only. Chemical identifiers include CAS 1457983-28-6, formula C25H25BrN6O2, and molecular weight 521.41 g/mol.
  • Potent ATP-competitive FLT3 inhibitor.
  • Supplied as a 10 mM solution in DMSO (1 mL).
  • Suitable for preclinical AML and resistance research.
  • High purity (98.0%).
  • Provided as the hydrochloride salt form.