Learn More
Creative Biomart Recombinant Dengue Type 1 Envelope Antigen, His-tagged
Supplier:  Creative Biomart DEN106
Encompass_Preferred
Recombinant Dengue Virus Subtype 1 Envelope 45 kDa (43-413a.a.) produced in E. coli, contains important epitopes for dengue antibody recognition. This protein is fused to a 6 His tag.
Catalog No. 50-208-4484