Learn More
Creative Biomart Recombinant Dengue Type 2 Envelope Antigen, His-tagged
Supplier:  Creative Biomart DEN111
Encompass_Preferred
Recombinant Dengue Virus Subtype 2 Envelope 32 kDa, produced in E. coli, amino acids 45-297 and fused to a 6 His tag, containing domains I+II of the dengue envelope.
Catalog No. 50-208-4489