Learn More
Creative Biomart Recombinant Dengue NS1 Antigen, His-tagged
Supplier:  Creative Biomart DNS124
Encompass_Preferred
The E. coli derived recombinant protein contains the NS1 Dengue Virus full length Type-2 immunodominant regions, having an Mw of 45 kDa.The dengue protein is fused to 6 His tag at C-terminus and pu
Catalog No. 50-208-4502