Learn More
Enzo Life Sciences Dengue NS1 ST1, (recombinant) (100µg)
SDP
Catalog No. 502013899
Encompass_Preferred
Alternative name: DENV1-NS1. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In DPBS pH 7.4. Source: Produced in HEK 293 cells. Dengue Viral NS1 Serotype 1 (DENV1-NS1) from strain Nauru/Western Pacific/1974. Long Term Storage: -80°C. Applications: ELISA Vaccine research, LINE assay, Antibody capture.
Catalog No. 50-201-3899 Supplier Enzo Life Sciences Supplier No. ENZPRT1040100
Alternative name: DENV1-NS1. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In DPBS pH 7.4. Source: Produced in HEK 293 cells. Dengue Viral NS1 Serotype 1 (DENV1-NS1) from strain Nauru/Western Pacific/1974. Long Term Storage: -80°C. Applications: ELISA Vaccine research, LINE assay, Antibody capture.