Learn More
Creative Biomart Recombinant Dengue Subtype 3 NS1 Antigen, His-tagged
Supplier:  Creative Biomart DNS132
Encompass_Preferred
The E. coli derived recombinant protein contains the NS1 Dengue Virus full length Type-3 immunodominant regions, migrates as 45 kDa on 12% SDS-PAGE gel. The dengue protein is fused to a 6 His tag a
Catalog No. 50-208-4510