Learn More
Creative Biomart Recombinant Dengue Subtype 3 NS1 Antigen
Supplier:  Creative Biomart DNS129
Encompass_Preferred
Recombinant Dengue Virus NS1 Subtype 3 produced in insect cells is a polypeptide chain containing amino acids 775-1129 and having a molecular weight of approximately 50 kDa.Dengue NS1 ST3 is purified
Catalog No. 50-208-4507