Learn More
Bioss Dengue virus
SDP
Supplier:  Bioss BSM72108M
Encompass_Preferred
Dengue virus Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-65850