Learn More
Drg International Inc DRG Dengue Virus IgM ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3471R
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgM-class antibodies to Dengue virus in serum. This product is for Research Use Only. The Dengue Virus IgM ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Samples are diluted with Sample Diluent and additionally incubated with IgG-RF-Sorbent, containing hyper-immune anti-human IgG-class antibody to eliminate competitive inhibition from specific IgG and to remove rheumatoid factors. This pretreatment avoids false results. Microtiter wells as a solid phase are coated with Dengue - 2 Virus antigen.
Catalog No. 50-194-7567