Learn More
ACROBiosystems DENV2 (strain Thailand/16681/1984) NS1 protein, His Tag (NS1-D52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 776 - Ala 1127 (Accession # P29990).
Catalog No. 502253533
Encompass_Preferred
Dengue is a mosquito-borne viral disease widely spread all over the world transmitted by 4 serotypes of dengue virus (DENV). The symptoms range from mild dengue fever to dengue hemorrhagic fever (DHF) and dengue shock syndrome (DSS) with high mortality. Currently, most of the studies on DENV vaccines emphasize on its envelope protein (i.e., Sanofi’s tetravalent dengue vaccine Dengvaxia). However, the fact pointing out that the vaccines with Envelope protein may could have negative effect on people never infected by DENV which makes the non-structural protein NS1 become another great target for vaccine research with its ability to avoid Antibody Dependent Enhancement.
This item is not returnable. View return policy
Catalog No. 50-225-3533 Supplier ACROBiosystems Supplier No. NS1D52H3100UG
Dengue is a mosquito-borne viral disease widely spread all over the world transmitted by 4 serotypes of dengue virus (DENV). The symptoms range from mild dengue fever to dengue hemorrhagic fever (DHF) and dengue shock syndrome (DSS) with high mortality. Currently, most of the studies on DENV vaccines emphasize on its envelope protein (i.e., Sanofi’s tetravalent dengue vaccine Dengvaxia). However, the fact pointing out that the vaccines with Envelope protein may could have negative effect on people never infected by DENV which makes the non-structural protein NS1 become another great target for vaccine research with its ability to avoid Antibody Dependent Enhancement.