Learn More
ATCC Dengue virus type 2; Strain: New Guinea C
Supplier:  ATCC VR1584
Encompass
Dengue virus type 2 strain New Guinea C is propagated in LLC-MK2 Derivative cells (ATCC CCL-7.1). This strain was isolated from the serum of a febrile patient in 1944 and was deposited by the University of Maryland School of Medicine, the Yale Arbovirus Research Unit, and the National Institute of Allergy and Infectious Diseases (NIAID). It has applications in vector-borne research.
Catalog No. 50-238-2948