Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Denhardt's solution (100X)

Catalog No. AAJ62416AD
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62416AD 50 mL
AAJ62416AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62416AD Supplier Thermo Scientific Chemicals Supplier No. J62416AD
Add to Cart

Denhardt's Solution is used as a blocking reagent for northern or southern blots to prevent probe binding on open membrane space and non-target nucleic acids. Denhardt's reagent is recommended for Northern hybridization, hybridization of RNA, single copy Southern hybridization' or hybridization of DNA, immobilized on nylon membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Denhardt's solution
MDL Number MFCD00717549
Concentration 100X
Solubility Information Soluble in water.
Quantity 50 mL
Physical Form Liquid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.