Learn More
Crescent Chemical Co Inc Denhardt's Solution, 50X Concentration, 10mL
SDP
Catalog No. 50248406
Encompass_Preferred
Denhardt's Solution, 50X Concentration, 10mL, Molecular Biology Grade, DNase/RNase not detected.Suitable for nucleic acid hybridization. Denhardt's solution is a mixture of blocking agents capable of saturating non-specific binding sites and to be used in membrane-based hybridization protocols. It is recommended for use with nylon membranes. Storage Conditions: -15 °C TO -25 °C
Catalog No. 50-248-406 Supplier Crescent Chemical Co Inc Supplier No. SE39603.01
Denhardt's Solution, 50X Concentration, 10mL, Molecular Biology Grade, DNase/RNase not detected.Suitable for nucleic acid hybridization. Denhardt's solution is a mixture of blocking agents capable of saturating non-specific binding sites and to be used in membrane-based hybridization protocols. It is recommended for use with nylon membranes. Storage Conditions: -15 °C TO -25 °C