Learn More
Bioworld Denhardt's Solution (100X), 10 mL
Catalog No. 50196818
Encompass_Preferred
Denhardt's Solution, 100X Concentrate Acts as a blocking reagent in hybridizations to help prevent probe binding to open membrane space and non-target nucleic acids. Specifications:Ultra Pure GradePurity: 99.0%pH @ 25°C: 5.0-7.0 Conductivity @ 25°C (µmhos): <350 Abs @ 278nm (5% water): 0.600-0.750 DNAse: None detectedRNAse: None detectedStorage: -15 to -22°C
Catalog No. 50-196-818 Supplier Bioworld Supplier No. 107500051
Add to Cart
Add to Cart
Denhardt's Solution, 100X Concentrate Acts as a blocking reagent in hybridizations to help prevent probe binding to open membrane space and non-target nucleic acids. Specifications:Ultra Pure GradePurity: 99.0%pH @ 25°C: 5.0-7.0 Conductivity @ 25°C (µmhos): <350 Abs @ 278nm (5% water): 0.600-0.750 DNAse: None detectedRNAse: None detectedStorage: -15 to -22°C