Learn More
Sigma Aldrich Fine Chemicals Biosciences Denhardts Solution 50x for
Catalog No. 502905597
Encompass
Denhardts SOLUTIONon is a blocking agent routinely used to reduce non-specific binding of detection reagents to the Northern and Southern Blot membranes. This 50x concentrate can be diluted to any concentration for use in a variety of pre-hybridization and hybridization buffers.
Catalog No. 50-290-5597 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D25325X5ML
Denhardts SOLUTIONon is a blocking agent routinely used to reduce non-specific binding of detection reagents to the Northern and Southern Blot membranes. This 50x concentrate can be diluted to any concentration for use in a variety of pre-hybridization and hybridization buffers.